Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tanks, rectangular, with cover, Nalgene®

Tanks, rectangular, with cover, Nalgene®

120 L, 610×457×450 mm
Catalog # NALG14200-0045
   Low Manufacturing Impact
Tanks, rectangular, with cover, Nalgene®
Catalog # NALG14200-0045
   Low Manufacturing Impact
Tanks, rectangular, with cover, Nalgene®
Tanks, rectangular, with cover, Nalgene®

120 L, 610×457×450 mm

Price
980,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These PP rectangular tanks with cover can significantly reduce evaporation and contamination.

  • Good stress-crack resistance
  • Suitable for use with many organic chemicals
  • Tanks can be modified with a factory-installed spigot
Specifications
  • Capacity:
    120 L
  • Seq:
    06
  • W×D×H:
    610×457×450 mm
  • Cat. No.:
    NALG14200-0045
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.