Orders LinkContactUsLinkComponent
Dressing forceps, Potts-Smith type
Dressing forceps, Potts-Smith type
Potts-Smith type, Straight, pointed, 180 mm
Catalog # MART12-192-18
Supplier:   KLS Martin Group
Dressing forceps, Potts-Smith type
Catalog # MART12-192-18
Supplier:  KLS Martin Group
Dressing forceps, Potts-Smith type
Potts-Smith type, Straight, pointed, 180 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel forceps.
  • Also available with TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life of the instruments
Specifications
  • Type:
    Straight, pointed
  • Description:
    Potts-Smith type
  • Length:
    180 mm