Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH-Fix pH Indicator Test Strips

pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax, 7,0 - 14,0, Classic flat box , 66×10×90 mm, 34,51 g
Catalog # 662-0207
Supplier:   MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
Catalog # 662-0207
Supplier:  MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax, 7,0 - 14,0, Classic flat box , 66×10×90 mm, 34,51 g

Price
16,10 €
Log in for availability
1 * 100 ST
Product Details & Documents

pH-Fix indicator test strips allow fast pH testing directly at the point of interest, and are suitable for untrained users as well as analytical professionals.

  • No accessories needed - always ready to use
  • Brilliant colour chart for precise readings
  • Long handle for sufficient hand - sample - distance
  • Non bleeding - no contamination of sample solutions

The indicator dyes used for pH-Fix test strips are chemically bound to the test pads to prevent bleeding and, therefore, contamination of the sample. Instructions for use according to IVD Directive 98/79/EC - The instructions for use according to IVD Directive 98/79/EC can be requested by providing the article number and LOT number at the email address [email protected].

Caution: No hazardous material.

Specifications
  • pH range:
    7,0 - 14,0
  • Description:
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
  • Seq:
    15
  • Pack type:
    Classic flat box , 66×10×90 mm, 34,51 g
  • Cat. No.:
    662-0207
  • Shelf life:
    4 Year(s)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.