Order Entry
Belgium
Orders LinkContactUsLinkComponent
Test Strips, Peroxide

Test Strips, Peroxide

QUANTOFIX® peroxide 100, 0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂; 1 - 100 - mg/L - H₂O₂, Redox reaction (peroxidase/redox indicator), 43×126×43 mm (packaging), 100 Tests, 100 Tests
Catalog # 705-0313
Supplier:   MACHEREY-NAGEL
Test Strips, Peroxide
Catalog # 705-0313
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test Strips, Peroxide
Test Strips, Peroxide

QUANTOFIX® peroxide 100, 0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂; 1 - 100 - mg/L - H₂O₂, Redox reaction (peroxidase/redox indicator), 43×126×43 mm (packaging), 100 Tests, 100 Tests

CAS Number:  
Price
40,38 €
Log in for availability
1 * 1 EA
Product Details & Documents

QUANTOFIX Peroxide test strips enables the quick and reliable determination of peroxides in water–based liquids. The simple handling makes the test strip particularly user–friendly: simply dip it into the sample, wait briefly and read off the result – in less than a minute.

  • Peroxide - Redox reaction (white → blue or brown for 705-1154)

The test strip can be evaluated with the included colour scale as well as instrumental with the QUANTOFIX Relax strip reader. In addition to a convenient measuring procedure, this also offers automated and digital documentation of the results. QUANTOFIX Peroxide is ideal for the food industry. The test strips can be used to check rinsing and washing processes of equipment quickly, easily and reliably – disinfectant residues in bottles and yoghurt pots can be easily identified. The same applies to medical devices. QUANTOFIX Peroxide  is an ideal companion for food quality and patient safety.

Specifications

Specifications

  • Chemical method:
    Redox reaction (peroxidase/redox indicator)
  • Pk:
    100 Tests
  • Description:
    QUANTOFIX® peroxide 100
  • Dimensions:
    43×126×43 mm (packaging)
  • Tests per kit:
    100 Tests
  • Range:
    0 • 1 • 3 • 10 • 30 • 100 - mg/L - H₂O₂; 1 - 100 - mg/L - H₂O₂
  • Cat. No.:
    705-0313
  • Parameters:
    All reagents included
  • Contents:
    100 test strips in a tube, instruction leaflet
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    QUANTOFIX Relax
  • Shelf life (months):
    30 (from production)
  • Method:
    Peroxide - Redox reaction (white → blue)
  • Weight:
    49 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.