Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Papers, without Colour Scale

pH Papers, without Colour Scale

pH test paper Litmus paper (red → violet → blue) blue, refill pack, 3 reels, length 5 m and width 7 mm, 8,0 - 5,0, 60×60×33 mm, 19,8 g
Catalog # MANA91116
Supplier:   MACHEREY-NAGEL
pH Papers, without Colour Scale
Catalog # MANA91116
Supplier:  MACHEREY-NAGEL
pH Papers, without Colour Scale
pH Papers, without Colour Scale

pH test paper Litmus paper (red → violet → blue) blue, refill pack, 3 reels, length 5 m and width 7 mm, 8,0 - 5,0, 60×60×33 mm, 19,8 g

Price
17,40 €
Log in for availability
1 * 3 ROU
Product Details & Documents

Easy to use pH papers to control pH adjustments.

  • Long term stable and protected against external influences due to plastic packaging

  • No accessories needed - always ready to use
  • Quick and easy determination of the pH

These products are simple, completely impregnated indicator papers, which do not allow exact pH determinations, but pH adjustments during special chemical procedures. For this reason, no colour scale is provided with these papers.

Certifications: Not CE certified.

Caution: Contains hazardous materials: MANA90703 and MANA90705.

Specifications
  • pH range:
    8,0 - 5,0
  • Description:
    pH test paper Litmus paper (red → violet → blue) blue, refill pack, 3 reels, length 5 m and width 7 mm
  • Seq:
    9
  • Pack type:
    60×60×33 mm, 19,8 g
  • Cat. No.:
    MANA91116
  • Shelf life:
    3 year(s)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.