pH Papers, without Colour Scale
Product Details & Documents
Easy to use pH papers to control pH adjustments.
Long term stable and protected against external influences due to plastic packaging
- No accessories needed - always ready to use
- Quick and easy determination of the pH
These products are simple, completely impregnated indicator papers, which do not allow exact pH determinations, but pH adjustments during special chemical procedures. For this reason, no colour scale is provided with these papers.
Certifications: Not CE certified.
Caution: Contains hazardous materials: MANA90703 and MANA90705.
Documents
Specifications
- pH range:8,0 - 5,0
- Description:pH test paper Litmus paper (blue → red) blue, 1 reel, length 5 m and width 7 mm
- Seq:4
- Pack type:63×17×63 mm, 20,5 g
- Cat. No.:MANA91106
- Shelf life:3 year(s)
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



