Dissecting scissors, straight
Straight, sharp, 125 mm
N° de catalogue 233-0012
Fournisseur: MURRAY S
Jump to:
Product Details & Documents
Stainless steel.
- Closed or open shanks
- Straight, sharp/sharp
Specifications
- Type:Straight, sharp
- Length:125 mm
- Cat. No.:233-0012