Order Entry
Belgium
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Incident halogen bulb, 12 V, 10 W
Catalog # 632-8943
Supplier:   MOTIC
Halogen bulbs
Catalog # 632-8943
Supplier:  MOTIC
Halogen bulbs
Halogen bulbs

Incident halogen bulb, 12 V, 10 W

Price
12,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Incident halogen bulb
  • Seq:
    03
  • Power:
    10 W
  • Voltage:
    12 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.