Orders LinkContactUsLinkComponent
Slide boxes, PS
Slide boxes, PS
Slide boxes, PS, 50, 89×33×208 mm
Catalog # 631-0052
Supplier:   LP ITALIANA
Slide boxes, PS
Catalog # 631-0052
Supplier:  LP ITALIANA
Slide boxes, PS
Slide boxes, PS
Slide boxes, PS, 50, 89×33×208 mm
Price
151,00 €
Log in for availability
1 * 24 ST
Product Details & Documents
Microscope slide boxes for 25 or 50 slides.
Specifications
  • No. of slides:
    50
  • Description:
    Slide boxes
  • Material:
    PS
  • Seq:
    02
  • W×D×H:
    89×33×208 mm