Order Entry
Belgium
Orders LinkContactUsLinkComponent
Swabs

Swabs

Sterile swabs (single wrapped), Aluminium/cotton, 1 mm, 150 mm
Catalog # LPIT112698
Supplier:   LP ITALIANA
Swabs
Catalog # LPIT112698
Supplier:  LP ITALIANA
Swabs
Swabs

Sterile swabs (single wrapped), Aluminium/cotton, 1 mm, 150 mm

Price
528,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Sterile and non sterile swabs designed for use in microbiology.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Ø:
    1 mm
  • Description:
    Sterile swabs (single wrapped)
  • Length:
    150 mm
  • Shaft/head:
    Aluminium/cotton
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.