Orders LinkContactUsLinkComponent
Vacuum filter holders
Vacuum filter holders
130 ml, 47 mm
Catalog # SART16523
Supplier:   Sartorius
Vacuum filter holders
Catalog # SART16523
Supplier:  Sartorius
Vacuum filter holders
130 ml, 47 mm
Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These vacuum filter holders feature a glass cylinder, PP base and sealing plug with anodised aluminum closing cap.
  • Vacuum operated
Specifications
  • For Ø:
    47 mm
  • Capacity:
    130 ml
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Aluminium closing cap
    Aluminium closing cap
    SART6980591
    Log in for availability
    1 * 1 ST - 8,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Closing ring
    Closing ring
    SART6980590
    Log in for availability
    1 * 1 ST - 11,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass cylinder, 130 ml
    Glass cylinder, 130 ml
    SART6985060
    Log in for availability
    1 * 100 ST - 77,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass cylinder, 85 ml
    Glass cylinder, 85 ml
    SART6980589
    Log in for availability
    1 * 1 ST - 61,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Plug
    Plug
    SART6980588
    Log in for availability
    1 * 10 ST - 39,90 €
Product Information
  • Catalog #
    Availability
    Price
  • PP base
    PP base
    SART6980586
    Log in for availability
    1 * 1 ST - 11,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Flat silicone gasket
    Flat silicone gasket
    SART6980573
    Log in for availability
    1 * 1 ST - 6,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Silicone O-ring
    Silicone O-ring
    SART6980574
    Log in for availability
    1 * 1 ST - 8,50 €