Gas sampling tubes
500 ml, without threaded bleed connection
201-9113
Duran® clear borosilicate glass.
- Excellent thermal and chemical resistance
- Connection on one side with capillary tube (internal Ø: 2,7 mm, external Ø: 8,0 mm) and on the other side with tube connector (external Ø: 8,0 mm)
- With white labelling area
Gas collection tubes for removing, collecting and transporting gas samples. Both sides with a 3 NS/14 standard ground one-way stop cock.
: DIN 12473-1
- Type:without threaded bleed connection
- Capacity:500 ml