Orders LinkContactUsLinkComponent
Soxhlet extractors
Soxhlet extractors
Extractor, 100 ml, 45/40 NS
Catalog # 533-2143
Supplier:   Lenz Laborglas
Soxhlet extractors
Catalog # 533-2143
Supplier:  Lenz Laborglas
Soxhlet extractors
Soxhlet extractors
Extractor, 100 ml, 45/40 NS
Price
118,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass. For extraction from solids with solvents.
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Delivery information: The complete apparatus consists of a round bottom flask with NS 29/32 socket, an extractor with NS 29/32 cone and a condenser with GL 14 tubing connectors. All parts are available separately.
Documents
Specifications
  • Extraction cap.:
    100 ml
  • Socket size:
    45/40 NS
  • Type:
    Extractor
  • Seq:
    18