Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Soxhlet extractors

Complete, 200 ml, 250 ml, 45/40 NS, 45/40
  533-2135
Soxhlet extractors
  533-2135
Soxhlet extractors
Soxhlet extractors

Complete, 200 ml, 250 ml, 45/40 NS, 45/40

 
317,00 €
   
1 * 1 ST
 

DURAN®, borosilicate glass. For extraction from solids with solvents.

The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.

 : The complete apparatus consists of a round bottom flask with NS 29/32 socket, an extractor with NS 29/32 cone and a condenser with GL 14 tubing connectors. All parts are available separately.

 
  • Extraction cap.:
    200 ml
  • NS:
    45/40
  • Socket size:
    45/40 NS
  • Type:
    Complete
  • Seq:
    05
  • Flask capacity:
    250 ml
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.