Orders LinkContactUsLinkComponent
Funnels with conical joint
Funnels with conical joint
24/29 NS, 70 mm
Catalog # 221-0137
Supplier:   Lenz Laborglas
Funnels with conical joint
Catalog # 221-0137
Supplier:  Lenz Laborglas
Funnels with conical joint
Funnels with conical joint
24/29 NS, 70 mm
Price
40,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.
Documents
Specifications
  • Cone size:
    24/29 NS
  • Funnel O-Ø:
    70 mm