Adapters, delivery, bent
19/26 NS, 19/26 NS
Catalog # 201-3109
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With core and jacket. Inclined at an angle of 105°. Cone with drip point.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:19/26 NS
- Socket size:19/26 NS