Order Entry
Belgium
Orders LinkContactUsLinkComponent
Adapters, delivery, angled

Adapters, delivery, angled

14/23 NS, 190 mm
Catalog # 201-0365
Supplier:   Lenz Laborglas
Adapters, delivery, angled
Catalog # 201-0365
Supplier:  Lenz Laborglas
Adapters, delivery, angled
Adapters, delivery, angled

14/23 NS, 190 mm

Price
22,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate 3.3 glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.

Documents

Specifications
  • Socket size:
    14/23 NS
  • Tube length:
    190 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.