Adapters, delivery, angled
29/32 NS, 65 mm
Catalog # 201-3108
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Socket size:29/32 NS
- Tube length:65 mm