Orders LinkContactUsLinkComponent
Distillation columns, Vigreux
Distillation columns, Vigreux
600 mm, 24/29 NS, 24/29 NS
Catalog # 201-1001
Supplier:   Lenz Laborglas
Distillation columns, Vigreux
Catalog # 201-1001
Supplier:  Lenz Laborglas
Distillation columns, Vigreux
Distillation columns, Vigreux
600 mm, 24/29 NS, 24/29 NS
Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air cooled fractionating column with alternating horizontal and inclined indentations.
Documents
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Usable length:
    600 mm
  • Seq:
    05