Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
14/23 NS, 14/23 NS
Catalog # 271-2221
Supplier:   Lenz Laborglas
Multiple adapters
Catalog # 271-2221
Supplier:  Lenz Laborglas
Multiple adapters
Multiple adapters
14/23 NS, 14/23 NS
Price
44,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
Documents
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS