Orders LinkContactUsLinkComponent
Stopcocks, three way
Stopcocks, three way
1,5 mm, 14,5 NS
N° de catalogue 201-1109
Fournisseur:   Lenz Laborglas
Stopcocks, three way
N° de catalogue 201-1109
Fournisseur:  Lenz Laborglas
Stopcocks, three way
Stopcocks, three way
1,5 mm, 14,5 NS
Prix
51,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
Specifications
  • Hole Ø:
    1,5 mm
  • Spigot:
    14,5 NS