Orders LinkContactUsLinkComponent
Stopcocks, three way
Stopcocks, three way
6,0 mm, 29,2 NS
Catalog # 227-3235
Supplier:   Lenz Laborglas
Stopcocks, three way
Catalog # 227-3235
Supplier:  Lenz Laborglas
Stopcocks, three way
Stopcocks, three way
6,0 mm, 29,2 NS
Price
93,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
Specifications
  • Hole Ø:
    6,0 mm
  • Spigot:
    29,2 NS