Orders LinkContactUsLinkComponent
Stopcocks, three way
4,0 mm, 18,8 NS
N° de catalogue 201-1036
Fournisseur:   Lenz Laborglas
Stopcocks, three way
N° de catalogue 201-1036
Fournisseur:  Lenz Laborglas
Stopcocks, three way
Stopcocks, three way
4,0 mm, 18,8 NS
Prix
58,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
Specifications
  • Hole Ø:
    4,0 mm
  • Spigot:
    18,8 NS