Stopcocks, three way
2,5 mm, 14,5 NS
Catalog # 201-1035
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys with T-bore
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Hole Ø:2,5 mm
- Spigot:14,5 NS