Stopcocks, two-way
2,5 mm, 14,5 NS
Catalog # 227-3123
Supplier: Lenz Laborglas
Aller à :
Product Details & Documents
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
- Excellent chemical resistance
- Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Spécifications
- Hole Ø:2,5 mm
- Spigot:14,5 NS