Orders LinkContactUsLinkComponent
Stopcocks, one-way
Stopcocks, one-way
1,5 mm, 12,5 NS
Catalog # 201-1106
Supplier:   Lenz Laborglas
Stopcocks, one-way
Catalog # 201-1106
Supplier:  Lenz Laborglas
Stopcocks, one-way
Stopcocks, one-way
1,5 mm, 12,5 NS
Price
30,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
Specifications
  • Hole Ø:
    1,5 mm
  • Seq:
    11
  • Spigot:
    12,5 NS