Orders LinkContactUsLinkComponent
Spherical ground joints
Spherical ground joints
120 mm, S 29, -
Catalog # 201-4224
Supplier:   Lenz Laborglas
Spherical ground joints
Catalog # 201-4224
Supplier:  Lenz Laborglas
Spherical ground joints
Spherical ground joints
120 mm, S 29, -
Price
14,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
Spherical cup and ball joint with inch markings. Fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641
Certifications: DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641.
Documents
Specifications
  • Ball:
    S 29
  • Socket:
    -
  • Total length:
    120 mm