Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

Kryo 30, –30...+90 °C, − °C, − °C, Light yellow, 20 l
Catalog # 461-2876
Supplier:   LAUDA
Bath fluids
Catalog # 461-2876
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Kryo 30, –30...+90 °C, − °C, − °C, Light yellow, 20 l

CAS Number:  
Price
674,00 €
Log in for availability
1 * 20 L
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    20 l
  • Colour:
    Light yellow
  • Fire point:
    − °C
  • Flash point:
    − °C
  • Temperature range:
    –30...+90 °C
  • Type:
    Kryo 30

Specifications