Orders LinkContactUsLinkComponent
Decontamination solution, DNA AWAY®
DNA AWAY®, 250 mL
  732-2353
   Low Manufacturing Impact
Decontamination solution, DNA AWAY®
  732-2353
   Low Manufacturing Impact
 :  
Decontamination solution, DNA AWAY®
Decontamination solution, DNA AWAY®
DNA AWAY®, 250 mL
 :  
22,00 €
  30/06/2026
1 * 250 mL
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
  • Suitable for PCR applications, preventing false-positive PCR results
  • Easy cleaning of various types of surfaces
  • Pk:
    250 mL
  • Description:
    DNA AWAY®
  • Cat. No.:
    732-2353
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.