Orders LinkContactUsLinkComponent
Decontamination solution, RNase AWAY®
Decontamination solution, RNase AWAY®
RNase AWAY®, 1 L
Catalog # 732-2271
   Low Manufacturing Impact
Decontamination solution, RNase AWAY®
Catalog # 732-2271
   Low Manufacturing Impact
CAS Number:  
Decontamination solution, RNase AWAY®
Decontamination solution, RNase AWAY®
RNase AWAY®, 1 L
CAS Number:  
Price
55,00 €
Sale
Valid until 30/06/2026
Log in for availability
1 * 1 L
Product Details & Documents
Ready-to-use solution for fast, easy elimination of RNase contamination from work surfaces, instruments, pipettes, glass and consumables.
  • Easy cleaning of various types of surfaces
  • Ideal for use on materials that are not autoclavable
  • Replaces time-consuming processes such as DEPC treatment
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers, etc.
Specifications
Specifications
  • Pk:
    1 L
  • Description:
    RNase AWAY®
  • Cat. No.:
    732-2271
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Specifications