Decontamination solution, RNase AWAY®
RNase AWAY®, 250 mL
Supplier: Thermo Fisher Scientific
Decontamination solution, RNase AWAY®
Catalog # 732-2270
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Ready-to-use solution for fast, easy elimination of RNase contamination from work surfaces, instruments, pipettes, glass and consumables.
- Easy cleaning of various types of surfaces
- Ideal for use on materials that are not autoclavable
- Replaces time-consuming processes such as DEPC treatment
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers, etc.
Specifications
Specifications
- Pk:250 mL
- Description:RNase AWAY®
- Cat. No.:732-2270
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.