Dewar flasks, type GSS
Loose lid insulating plug, GSS 1000
Catalog # 478-5143
Supplier: KGW
Jump to:
Product Details & Documents
Dewar flasks in stainless steel for storage and transport of liquid nitrogen, dry ice and other coolants. They are vacuum insulated and have a temperature range from −200 to +200 °C.
- With side grip and transport handle (except model GSS 500)
Specifications
- Description:Loose lid insulating plug
- Seq:08
- For:GSS 1000