Spherical Dewar flasks, type AL
Glass refill, 22 A, Spherical dewar flask, 22 AL
Catalog # 478-0735
Supplier: KGW
Jump to:
Product Details & Documents
Dewar flasks made from silvered borosilicate glass, brushed aluminium protective covering.
- With lid and handle
Specifications
- Description:Glass refill, 22 A
- Seq:05
- For:Spherical dewar flask, 22 AL