Order Entry
Belgium
Orders LinkContactUsLinkComponent
Scintillation vials

Scintillation vials

4,00 ml, 13,7×53,1 mm, Yes
Catalog # 216-4309
Supplier:   KARTELL
Scintillation vials
Catalog # 216-4309
Supplier:  KARTELL
Scintillation vials
Scintillation vials

4,00 ml, 13,7×53,1 mm, Yes

Price
289,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents

HDPE, translucent. Compatible with all scintillation counters. A special screw cap ensures leakproof handling.

Documents

Specifications
  • Cap included:
    Yes
  • Ø×H:
    13,7×53,1 mm
  • Capacity:
    4,00 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.