Scintillation vials
20,00 ml, 26,5×58,5 mm, Yes
Catalog # 216-4308
Supplier: KARTELL
Jump to:
Product Details & Documents
HDPE, translucent. Compatible with all scintillation counters. A special screw cap ensures leakproof handling.
Documents
KART933_TDS_EN
Datasheet
Specifications
- Cap included:Yes
- Ø×H:26,5×58,5 mm
- Capacity:20,00 ml
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



