Orders LinkContactUsLinkComponent
Filter discs
Filter discs
160 mm
Catalog # 519-5007
Supplier:   KARTELL
Filter discs
Catalog # 519-5007
Supplier:  KARTELL
Filter discs
Filter discs
160 mm
Price
19,30 €
Log in for availability
1 * 10 ST
Product Details & Documents
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
Specifications
  • Ø:
    160 mm
  • Seq:
    07