Order Entry
Belgium
Orders LinkContactUsLinkComponent
Press-down stoppers

Press-down stoppers

Natural, 28 mm, 30 mm
N° de catalogue KART656
Fournisseur:   KARTELL
Press-down stoppers
N° de catalogue KART656
Fournisseur:  KARTELL
Press-down stoppers
Press-down stoppers

Natural, 28 mm, 30 mm

Prix
22,10 €
Log in for availability
1 * 100 ST
Product Details & Documents

For sealing glass or plastic reagent and sample tubes.

  • PE, natural colour
  • Good chemical resistance
  • Economical
  • Convenient pack sizes

*conical

Specifications
  • Ø bottom:
    28 mm
  • Ø top:
    30 mm
  • Colour:
    Natural