Order Entry
Belgium
Orders LinkContactUsLinkComponent
Press-down stoppers

Press-down stoppers

Natural, 14,5 mm, 16 mm
Catalog # 212-0214
Supplier:   KARTELL
Press-down stoppers
Catalog # 212-0214
Supplier:  KARTELL
Press-down stoppers
Press-down stoppers

Natural, 14,5 mm, 16 mm

Price
9,60 €
Log in for availability
1 * 100 ST
Product Details & Documents

For sealing glass or plastic reagent and sample tubes.

  • PE, natural colour
  • Good chemical resistance
  • Economical
  • Convenient pack sizes

*conical

Specifications
  • Ø bottom:
    14,5 mm
  • Ø top:
    16 mm
  • Colour:
    Natural
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.