Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Laboratory trays

202×151×81 mm
  216-2109
 :   KARTELL
Laboratory trays
  216-2109
 :  KARTELL
Laboratory trays
Laboratory trays

202×151×81 mm

 
15,30 €
   
1 * 1 ST
 

PS, white.

  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning

General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.

 
  • Seq:
    03
  • W×D×H:
    202×151×81 mm