Order Entry
Belgium
Orders LinkContactUsLinkComponent
Laboratory trays

Laboratory trays

252×252×42 mm
Catalog # 216-2108
Supplier:   KARTELL
Laboratory trays
Catalog # 216-2108
Supplier:  KARTELL
Laboratory trays
Laboratory trays

252×252×42 mm

Price
18,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

PS, white.

  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning

General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.

Documents

Specifications
  • Seq:
    04
  • W×D×H:
    252×252×42 mm