Order Entry
Belgium
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

810 ml, 110 mm
Catalog # 519-2365
Supplier:   KARTELL
Büchner funnels
Catalog # 519-2365
Supplier:  KARTELL
Büchner funnels
Büchner funnels

810 ml, 110 mm

Price
48,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white.

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    810 ml
  • For filter Ø:
    110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.