Order Entry
Belgium
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

390 ml, 90 mm
Catalog # 519-2364
Supplier:   KARTELL
Büchner funnels
Catalog # 519-2364
Supplier:  KARTELL
Büchner funnels
Büchner funnels

390 ml, 90 mm

Price
35,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white.

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    390 ml
  • For filter Ø:
    90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.