Order Entry
Belgium
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

70 ml, 55 mm
Catalog # 519-2362
Supplier:   KARTELL
Büchner funnels
Catalog # 519-2362
Supplier:  KARTELL
Büchner funnels
Büchner funnels

70 ml, 55 mm

Price
20,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white.

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    70 ml
  • For filter Ø:
    55 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.