Funnels with short stem
360 mm, 420 mm
Catalog # 221-0082
Supplier: KARTELL
Jump to:
Product Details & Documents
HDPE, translucent.
- Good chemical resistance
- With raised lip to prevent overflowing
- External ribs for unobstructed removal of air when filtering
With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Documents
KART175_TDS_EN
Datasheet
Specifications
- Funnel O-Ø:420 mm
- Length:360 mm
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



