Weighing bottles, PP
23 ml, 30×48 mm
Catalog # 611-0103
Supplier: KARTELL
Jump to:
Product Details & Documents
PP with lid.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Documents
Specifications
- Ø×H:30×48 mm
- Capacity:23 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



