Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tanks

Tanks

Grey, 75 l, 457×750×310 mm
Catalog # 216-0258
Supplier:   KARTELL
Tanks
Catalog # 216-0258
Supplier:  KARTELL
Tanks
Tanks

Grey, 75 l, 457×750×310 mm

Price
197,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of HDPE, grey.

  • Moulded grips
  • Sturdy

Useful general purpose tanks that can be stacked for storage.

Specifications
  • Capacity:
    75 l
  • Colour:
    Grey
  • W×D×H:
    457×750×310 mm