Orders LinkContactUsLinkComponent
Drip closures
Drip closures
Screw cap with 19 mm nozzle, GL 25, Short spout dropping bottles
Catalog # 217-0489
Supplier:   KARTELL
Drip closures
Catalog # 217-0489
Supplier:  KARTELL
Drip closures
Drip closures
Screw cap with 19 mm nozzle, GL 25, Short spout dropping bottles
Price
1,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
HDPE, natural colour.
  • Good chemical resistance, good temperature resistance
  • Available in two lengths
  • Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.
Specifications
  • For:
    Short spout dropping bottles
  • Thread:
    GL 25
  • Description:
    Screw cap with 19 mm nozzle
  • Seq:
    03