Polyethylene threaded water bottle caps
50/100 ml, 215-1329/215-1330, GL 18
Catalog # KART3300
Supplier: KARTELL
Jump to:
Product Details & Documents
LDPE.
- Fine tip tube
Specifications
- For:215-1329/215-1330
- Thread:GL 18
- Seq:01
- For bottles:50/100 ml