Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Slide boxes

Grey, PS, 25, 83×98×38 mm
  631-9140
 :   KARTELL
Slide boxes
  631-9140
 :  KARTELL
Slide boxes
Slide boxes

Grey, PS, 25, 83×98×38 mm

 
21,70 €
   
1 * 1 ST
 

High impact resistant slide boxes with removable lids.

  • Well spaced slots for easy handling of slides
  • Numbering for cross reference with corresponding index card
  • Locating lugs so boxes can be stacked

W×D×H: 83×98×38 mm (for 25); 97×230×35 mm (for 50); 280×230×35 mm (for 100)

 
  • No. of slides:
    25
  • Colour:
    Grey
  • Material:
    PS
  • W×D×H:
    83×98×38 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.