Order Entry
Belgium
Orders LinkContactUsLinkComponent
Funnel holders

Funnel holders

2 funnels
Catalog # 221-1262
Supplier:   KARTELL
Funnel holders
Catalog # 221-1262
Supplier:  KARTELL
Funnel holders
Funnel holders

2 funnels

Price
43,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, white, unbreakable.

  • For funnels 50 to 120 mm Ø
  • With bosshead to fit rods 8 to 14 mm Ø
  • Interchangeable insert for different sizes
Specifications
  • For:
    2 funnels
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.